Gene Transcripts

Gene Symbol
OR1B1
Entrez Gene ID
347169
Gene Name
olfactory receptor family 1 subfamily B member 1 (gene/pseudogene)
Gene Aliases
OR9-26, OR9-B
Chromosome Location
Chr. 9: 122578405 - 122657626 on Build GRCh38
UniGene ID
Hs.553710
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004450.1 NP_001004450.1 1 763
XM_017014695.1 XP_016870184.1 2 1292
XM_017014696.1 XP_016870185.1 2 1324

View Full Product Details