Gene Transcripts

Gene Symbol
LY6D
Entrez Gene ID
8581
Gene Name
lymphocyte antigen 6 complex, locus D
Gene Aliases
E48, Ly-6D
Chromosome Location
Chr. 8: 142784882 - 142786592 on Build GRCh38
UniGene ID
Hs.415762
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003695.2 NP_003686.1 3 735
GenBank mRNA BC031330.1 - 3 735
BC034542.1 - 1 1356
X82693.1 - 3 671

View Full Product Details