Gene Transcripts

Gene Symbol
OR8J1
Entrez Gene ID
219477
Gene Name
olfactory receptor family 8 subfamily J member 1
Gene Aliases
OR11-183
Chromosome Location
Chr. 11: 56360215 - 56361288 on Build GRCh38
UniGene ID
Hs.626841
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005205.2 NP_001005205.2 1 235
GenBank mRNA BC137066.1 - 1 235
BC137067.1 - 1 235 9021064

View Full Product Details