Gene Transcripts

Gene Symbol
SUPT7L
Entrez Gene ID
9913
Gene Name
SPT7-like STAGA complex gamma subunit
Gene Aliases
SPT7L, STAF65, STAF65(gamma), STAF65G, SUPT7H
Chromosome Location
Chr. 2: 27642568 - 27663840 on Build GRCh38
UniGene ID
Hs.6232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282729.1 NP_001269658.1 6 1753
NM_001282730.1 NP_001269659.1 6 1961
NM_001282731.1 NP_001269660.1 6 1803
NM_001282732.1 NP_001269661.1 5 1343
NM_014860.2 NP_055675.1 6 1748
XM_005264672.4 XP_005264729.1 6 1950
GenBank mRNA AB018307.1 - 6 1490
AF197954.1 - 6 1509
AF224759.1 - 5 2338
AK000329.1 - 6 1518
AK000616.1 - 6 1304
AK304889.1 - 5 1126
BC074000.1 - 6 1490 6501232
CR456983.1 - 4 1147

View Full Product Details