Gene Transcripts

Gene Symbol
SH2D1B
Entrez Gene ID
117157
Gene Name
SH2 domain containing 1B
Gene Aliases
EAT2
Chromosome Location
Chr. 1: 162395266 - 162412138 on Build GRCh38
UniGene ID
Hs.350581
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053282.4 NP_444512.2 4 613
GenBank mRNA AK097162.1 - 4 540
BC022407.1 - 4 601 4717041
BC066595.1 - 3 422 5755414

View Full Product Details