Gene Transcripts

Gene Symbol
NKAPL
Entrez Gene ID
222698
Gene Name
NFKB activating protein like
Gene Aliases
C6orf194, bA424I5.1
Chromosome Location
Chr. 6: 28259297 - 28260958 on Build GRCh38
UniGene ID
Hs.48787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007531.2 NP_001007532.1 1 1507
GenBank mRNA BC038240.1 - 1 1437

View Full Product Details