Gene Transcripts

Gene Symbol
Col4a4
Entrez Gene ID
301562
Gene Name
collagen, type IV, alpha 4
Gene Aliases
Chromosome Location
Chr. 9: 88314763 - 88357183 on Build Rnor_6.0
UniGene ID
Rn.22075
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008332.1 NP_001008333.1 7 936
GenBank mRNA BC086403.1 - 7 936

View Full Product Details