Gene Transcripts

Gene Symbol
Vom1r40
Entrez Gene ID
494256
Gene Name
vomeronasal 1 receptor 40
Gene Aliases
V1rg1
Chromosome Location
Chr. 1: 67660337 - 67661329 on Build Rnor_6.0
UniGene ID
Rn.129426
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008904.1 NP_001008904.1 1 418
GenBank mRNA AY510277.1 - 1 418

Gene Symbol
LOC102551504
Entrez Gene ID
102551504
Gene Name
vomeronasal type-1 receptor 4-like
Gene Aliases
Chromosome Location
Chr. 1: 67917477 - 67918469 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006228159.1 XP_006228221.1 1 418

View Full Product Details