Gene Transcripts

Gene Symbol
Ly49i9
Entrez Gene ID
494205
Gene Name
Ly49 inhibitory receptor 9
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.160080
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009496.1 NP_001009496.1 5, 6 729
GenBank mRNA AY649839.1 - 5, 6 729

View Full Product Details