Gene Transcripts

Gene Symbol
RAB3GAP1
Entrez Gene ID
22930
Gene Name
RAB3 GTPase activating protein catalytic subunit 1
Gene Aliases
P130, RAB3GAP, RAB3GAP130, WARBM1
Chromosome Location
Chr. 2: 135052265 - 135176667 on Build GRCh38
UniGene ID
Hs.306327
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172435.1 NP_001165906.1 18 1981
NM_012233.2 NP_036365.1 18 1981
XM_011510823.2 XP_011509125.1 18 1981
XM_011510825.2 XP_011509127.1 18 1981
GenBank mRNA AB449877.1 - 18 1938
AK022494.1 - 18 1953
BC022977.1 - 5 625
BC036732.1 - 1 50
BC146809.1 - 20 2218 8860368
D31886.1 - 18 1940

View Full Product Details