Gene Transcripts

Gene Symbol
Rfc2
Entrez Gene ID
116468
Gene Name
replication factor C (activator 1) 2
Gene Aliases
Chromosome Location
Chr. 12: 25130375 - 25143480 on Build Rnor_6.0
UniGene ID
Rn.113319
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053786.1 NP_446238.1 6 494
GenBank mRNA AF208499.1 - 2 58
BC082110.1 - 6 494 7135071

View Full Product Details