Gene Transcripts

Gene Symbol
Rasl11b
Entrez Gene ID
305302
Gene Name
RAS-like family 11 member B
Gene Aliases
Chromosome Location
Chr. 14: 36550353 - 36554574 on Build Rnor_6.0
UniGene ID
Rn.36357
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002830.2 NP_001002830.1 4 863
GenBank mRNA BC083755.1 - 4 863

View Full Product Details