Gene Transcripts
- Gene Symbol
- Suclg2
- Entrez Gene ID
- 20917
- Gene Name
- succinate-Coenzyme A ligase, GDP-forming, beta subunit
- Gene Aliases
- AF171077, AW556404, D6Wsu120e, SCS-betaG
- Chromosome Location
- Chr. 6: 95473009 - 95718846 on Build GRCm38
- UniGene ID
- Mm.371585
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011507.3 |
NP_035637.2
|
2, 3 |
292 |
|
GenBank mRNA |
AF058956.1 |
- |
1, 2 |
134 |
|
AK042987.1 |
- |
2, 3 |
247 |
|
AK045176.1 |
- |
2, 3 |
245 |
|
AK156263.1 |
- |
2, 3 |
247 |
|
AK167136.1 |
- |
2, 3 |
247 |
|
AK167339.1 |
- |
2, 3 |
531 |
|
AK172561.1 |
- |
2, 3 |
247 |
|
AK172633.1 |
- |
2, 3 |
247 |
|
AK217215.1 |
- |
1, 2 |
132 |
|
BC054425.1 |
- |
1, 2 |
45 |
|
BC080781.1 |
- |
2, 3 |
247 |
30470296 |
BI985474.1 |
- |
2, 3 |
284 |
|
EU234009.1 |
- |
2, 3 |
199 |
|
View Full Product Details