Gene Transcripts

Gene Symbol
Lrrc8e
Entrez Gene ID
72267
Gene Name
leucine rich repeat containing 8 family, member E
Gene Aliases
1810049O03Rik, C87354
Chromosome Location
Chr. 8: 4224280 - 4237470 on Build GRCm38
UniGene ID
Mm.25479
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028175.2 NP_082451.2 3 1371
GenBank mRNA BC080783.1 - 3 1371

View Full Product Details