Gene Transcripts

Gene Symbol
Fancd2
Entrez Gene ID
312641
Gene Name
Fanconi anemia, complementation group D2
Gene Aliases
Chromosome Location
Chr. 4: 145489869 - 145551479 on Build Rnor_6.0
UniGene ID
Rn.203979
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001719.1 NP_001001719.1 10 845
GenBank mRNA AY621075.1 - 10 845

View Full Product Details