Gene Transcripts

Gene Symbol
Olr91
Entrez Gene ID
293232
Gene Name
olfactory receptor 91
Gene Aliases
ratchr1-161156092-161157039_ORF
Chromosome Location
Chr. 1: 168447533 - 168448480 on Build Rnor_6.0
UniGene ID
Rn.141896
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000139.1 NP_001000139.1 1 516

View Full Product Details