Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320808.1 | NP_001307737.1 | 8 | 2690 | |
NM_014873.2 | NP_055688.1 | 8 | 2101 | ||
XM_005273362.3 | XP_005273419.1 | 9 | 2249 | ||
XM_011510228.2 | XP_011508530.1 | 9 | 2138 | ||
XM_011510229.2 | XP_011508531.1 | 8 | 2021 | ||
XM_017003004.1 | XP_016858493.1 | 7 | 3253 | ||
GenBank mRNA | D86960.1 | - | 8 | 2101 |