Gene Transcripts

Gene Symbol
LYRM5
Entrez Gene ID
144363
Gene Name
LYR motif containing 5
Gene Aliases
Chromosome Location
Chr. 12: 25195216 - 25205051 on Build GRCh38
UniGene ID
Hs.209151
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK095164.1 - 2 2269
BC071769.1 - 3 702 4246777
BC100276.1 - 3 719 6178563

View Full Product Details