Gene Transcripts
- Gene Symbol
- OR4D2
- Entrez Gene ID
- 124538
- Gene Name
- olfactory receptor family 4 subfamily D member 2
- Gene Aliases
- BC2009, OR17-24
- Chromosome Location
- Chr. 17: 58169656 - 58170579 on Build GRCh38
- UniGene ID
- Hs.446879
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF303373.1 |
- |
1 |
1296 |
|
View Full Product Details