Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_212519.2 | NP_997684.1 | 2 | 548 | |
XM_008768643.1 | XP_008766865.1 | 2 | 842 | ||
XM_008768644.1 | XP_008766866.1 | 2 | 848 | ||
XM_008768645.1 | XP_008766867.1 | 2 | 846 | ||
GenBank mRNA | AB162856.1 | - | 2 | 411 | |
BC098640.1 | - | 2 | 548 | 7099109 | |
FQ228582.1 | - | 592 |