Gene Transcripts

Gene Symbol
Ring1
Entrez Gene ID
309626
Gene Name
ring finger protein 1
Gene Aliases
Ring1A
Chromosome Location
Chr. 20: 3812287 - 3815834 on Build Rnor_6.0
UniGene ID
Rn.116589
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212549.2 NP_997714.2 7 1537
GenBank mRNA BC098635.1 - 7 1481

View Full Product Details