Gene Transcripts

Gene Symbol
Nr1h5
Entrez Gene ID
381463
Gene Name
nuclear receptor subfamily 1, group H, member 5
Gene Aliases
Fxrb
Chromosome Location
Chr. 3: 102939658 - 102964290 on Build GRCm38
UniGene ID
Mm.331269
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198658.2 NP_941060.2 3 234
NM_201619.2 NP_963913.2 3 234
NM_201622.2 NP_963916.2 3 234
GenBank mRNA AY094586.1 - 3 234
AY094587.1 - 2 129
AY094588.1 - 2 129
AY094589.1 - 2 129
AY094590.1 - 2 129
AY094591.1 - 2 129
BC119524.1 - 2 129 40056073
BC119525.1 - 2 129 40056074

View Full Product Details