Gene Transcripts
- Gene Symbol
- POC1A
- Entrez Gene ID
- 25886
- Gene Name
- POC1 centriolar protein A
- Gene Aliases
- PIX2, SOFT, WDR51A
- Chromosome Location
- Chr. 3: 52075233 - 52154690 on Build GRCh38
- UniGene ID
- Hs.476306
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001161580.1 |
NP_001155052.1
|
8, 9 |
1194 |
|
NM_001161581.1 |
NP_001155053.1
|
8, 9 |
1265 |
|
NM_015426.4 |
NP_056241.3
|
8, 9 |
1194 |
|
XM_011533562.2 |
XP_011531864.1
|
8, 9 |
916 |
|
XM_011533565.2 |
XP_011531867.1
|
7, 8 |
782 |
|
XM_017006104.1 |
XP_016861593.1
|
8, 9 |
987 |
|
GenBank mRNA |
AK075289.1 |
- |
8, 9 |
909 |
|
AK315688.1 |
- |
8, 9 |
916 |
|
AL117629.1 |
- |
3, 4 |
233 |
|
BC007417.2 |
- |
7, 8 |
844 |
|
BC110877.1 |
- |
8, 9 |
1194 |
5588109 |
BC119692.2 |
- |
8, 9 |
904 |
40116721 |
BC119693.2 |
- |
8, 9 |
980 |
|
View Full Product Details