Gene Transcripts

Gene Symbol
CYP4F2
Entrez Gene ID
8529
Gene Name
cytochrome P450 family 4 subfamily F member 2
Gene Aliases
CPF2
Chromosome Location
Chr. 19: 15878024 - 15898120 on Build GRCh38
UniGene ID
Hs.558423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082.4 NP_001073.3 3 320
GenBank mRNA AK290790.1 - 3 274
AK296200.1 - 2 75
AK313705.1 - 3 265
BC067437.1 - 3 261 7002207
BC067438.1 - 3 261
BC067439.1 - 3 261 7002212
BC067440.1 - 3 261 7002214
D26480.1 - 3 265
DC362034.1 - 3 320
U02388.2 - 3 283

View Full Product Details