Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286643.1 | NP_001273572.1 | 6, 7 | 1028 | |
NM_015942.4 | NP_057026.3 | 6, 7 | 1028 | ||
XM_006716570.3 | XP_006716633.1 | 6, 7 | 986 | ||
XM_011517054.2 | XP_011515356.1 | 5, 6 | 695 | ||
GenBank mRNA | AF132946.1 | - | 5, 6 | 750 | |
AK001801.1 | - | 4, 5 | 732 | ||
AK309002.1 | - | 6, 7 | 1025 | ||
BC012995.2 | - | 6, 7 | 983 | 3529518 |