Gene Transcripts

Gene Symbol
INSIG2
Entrez Gene ID
51141
Gene Name
insulin induced gene 2
Gene Aliases
INSIG-2
Chromosome Location
Chr. 2: 118088418 - 118110031 on Build GRCh38
UniGene ID
Hs.7089
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321329.1 NP_001308258.1 5 861
NM_001321330.1 NP_001308259.1 4 486
NM_001321331.1 NP_001308260.1 5 494
NM_001321332.1 NP_001308261.1 5 487
NM_016133.3 NP_057217.2 5 868
GenBank mRNA AF125392.1 - 4 782
AF527632.1 - 5 817
AK291433.1 - 5 816
AK298599.1 - 4 450
BC022475.1 - 5 812 4794938
BE888848.1 - 5 493
BX647805.1 - 5 409
DC364733.1 - 4 451

View Full Product Details