Gene Transcripts
- Gene Symbol
- TRIM17
- Entrez Gene ID
- 51127
- Gene Name
- tripartite motif containing 17
- Gene Aliases
- RBCC, RNF16, terf
- Chromosome Location
- Chr. 1: 228407478 - 228416882 on Build GRCh38
- UniGene ID
- Hs.121748
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001024940.2 |
NP_001020111.1
|
5, 6 |
1136 |
|
NM_001134855.1 |
NP_001128327.1
|
5, 6 |
1288 |
|
NM_016102.3 |
NP_057186.1
|
5, 6 |
1172 |
|
XM_006711779.2 |
XP_006711842.1
|
5, 6 |
1199 |
|
XM_011544209.2 |
XP_011542511.1
|
5, 6 |
1186 |
|
XM_011544210.2 |
XP_011542512.1
|
5, 6 |
1288 |
|
XM_011544211.2 |
XP_011542513.1
|
5, 6 |
1189 |
|
GenBank mRNA |
AF156271.1 |
- |
5, 6 |
895 |
|
AK301105.1 |
- |
5, 6 |
1092 |
|
AK308774.1 |
- |
5, 6 |
1163 |
|
BC033788.1 |
- |
5, 6 |
936 |
5165609 |
View Full Product Details