Gene Transcripts

Gene Symbol
Taar5
Entrez Gene ID
294123
Gene Name
trace amine-associated receptor 5
Gene Aliases
taR-5
Chromosome Location
Chr. 1: 22547662 - 22548675 on Build Rnor_6.0
UniGene ID
Rn.140571
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009650.1 NP_001009650.1 1 454

View Full Product Details