Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012003.1 | NP_001012003.1 | 3 | 225 | |
XM_006244956.2 | XP_006245018.1 | 4 | 511 | ||
XM_008767081.1 | XP_008765303.1 | 3 | 1258 | ||
GenBank mRNA | AB164704.1 | - | 3 | 225 | |
BC129076.1 | - | 3 | 359 |