Gene Transcripts

Gene Symbol
Cpped1
Entrez Gene ID
302890
Gene Name
calcineurin-like phosphoesterase domain containing 1
Gene Aliases
RGD1306568
Chromosome Location
Chr. 10: 3655038 - 3774887 on Build Rnor_6.0
UniGene ID
Rn.79124
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013963.1 NP_001013985.1 4 1483
GenBank mRNA BC081991.1 - 4 1483
FQ219055.1 - 1 552

View Full Product Details