Gene Transcripts

Gene Symbol
Olr68
Entrez Gene ID
293214
Gene Name
olfactory receptor 68
Gene Aliases
ratchr1-160844789-160845757_ORF
Chromosome Location
Chr. 1: 168136959 - 168137927 on Build Rnor_6.0
UniGene ID
Rn.141915
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000133.1 NP_001000133.1 1 732

View Full Product Details