Gene Transcripts

Gene Symbol
Bud13
Entrez Gene ID
300687
Gene Name
BUD13 homolog (S. cerevisiae)
Gene Aliases
RGD1308076
Chromosome Location
Chr. 8: 50573026 - 50588860 on Build Rnor_6.0
UniGene ID
Rn.102777
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025277.1 NP_001020448.1 5 1123
GenBank mRNA BC097995.1 - 5 1123

View Full Product Details