Gene Transcripts

Gene Symbol
FAM83E
Entrez Gene ID
54854
Gene Name
family with sequence similarity 83 member E
Gene Aliases
Chromosome Location
Chr. 19: 48600600 - 48614841 on Build GRCh38
UniGene ID
Hs.165803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017708.3 NP_060178.2 5 1638
XM_005259019.1 XP_005259076.1 6 2962
GenBank mRNA AK000207.1 - 5 1764
CD722642.1 - 1 245

View Full Product Details