Gene Transcripts

Gene Symbol
OR10G3
Entrez Gene ID
26533
Gene Name
olfactory receptor family 10 subfamily G member 3
Gene Aliases
OR14-40
Chromosome Location
Chr. 14: 21569803 - 21570744 on Build GRCh38
UniGene ID
Hs.554581
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005465.1 NP_001005465.1 1 238

View Full Product Details