Gene Transcripts

Gene Symbol
MUC21
Entrez Gene ID
394263
Gene Name
mucin 21, cell surface associated
Gene Aliases
C6orf205, KMQK697, MUC-21
Chromosome Location
Chr. 6: 30983708 - 30988637 on Build GRCh38
UniGene ID
Hs.448277
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010909.3 NP_001010909.2 3 1905
NR_130720.1 3 2047
GenBank mRNA AB242595.1 - 3 1815
AK056612.1 - 2 400
AK298642.1 - 3 1893
AK301645.1 - 2 1613
AK316363.1 - 3 1895
BC105735.1 - 3 1840 40026480
BC105736.1 - 3 1982
BC105737.1 - 3 1840 40026476
BC107478.1 - 4 1390 40026469
HM596784.1 - 3 1834

View Full Product Details