Gene Transcripts

Gene Symbol
Nms
Entrez Gene ID
433292
Gene Name
neuromedin S
Gene Aliases
AB164466
Chromosome Location
Chr. 1: 38939149 - 38950276 on Build GRCm38
UniGene ID
Mm.333709
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011684.2 NP_001011684.3 10 926
GenBank mRNA AB164466.1 - 10 926

View Full Product Details