Gene Transcripts

Gene Symbol
Cmss1
Entrez Gene ID
288176
Gene Name
cms1 ribosomal small subunit homolog (yeast)
Gene Aliases
RGD1309437
Chromosome Location
Chr. 11: 45031697 - 45330481 on Build Rnor_6.0
UniGene ID
Rn.3487
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013866.1 NP_001013888.1 8, 9 787
GenBank mRNA BC089826.1 - 8, 9 787
FQ214930.1 - 8, 9 793
FQ219915.1 - 8, 9 788

View Full Product Details