Gene Transcripts

Gene Symbol
Slain1
Entrez Gene ID
361087
Gene Name
SLAIN motif family, member 1
Gene Aliases
RGD1308626
Chromosome Location
Chr. 15: 87862832 - 87906472 on Build Rnor_6.0
UniGene ID
Rn.21577
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014139.1 NP_001014161.1 6 1851
GenBank mRNA BC088867.1 - 6 1851

View Full Product Details