Gene Transcripts

Gene Symbol
Wdsub1
Entrez Gene ID
362137
Gene Name
WD repeat, sterile alpha motif and U-box domain containing 1
Gene Aliases
Chromosome Location
Chr. 3: 45924979 - 45955455 on Build Rnor_6.0
UniGene ID
Rn.101918
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014179.1 NP_001014201.1 11 1678
XM_006234208.2 XP_006234270.1 11 1938
XM_006234209.2 XP_006234271.1 11 1934
XM_006234211.2 XP_006234273.1 11 1659
GenBank mRNA BC089856.1 - 11 1678

View Full Product Details