Gene Transcripts

Gene Symbol
RMND1
Entrez Gene ID
55005
Gene Name
required for meiotic nuclear division 1 homolog
Gene Aliases
C6orf96, COXPD11, RMD1, bA351K16, bA351K16.3
Chromosome Location
Chr. 6: 151404548 - 151452181 on Build GRCh38
UniGene ID
Hs.486835
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017909.3 NP_060379.2 2 464
XM_017010988.1 XP_016866477.1 2 437
GenBank mRNA AK000634.1 - 2 407
AK292339.1 - 2 439
BC012081.1 - 1 301
BC106065.1 - 2 426 6165385
BC119683.1 - 2 329 40116343
DA747450.1 - 2 464

View Full Product Details