Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001009573.2 | NP_001009573.2 | 7 | 652 | |
GenBank mRNA | BC046519.1 | - | 7 | 652 | |
BC141289.1 | - | 6 | 564 | 9055901 | |
CF164660.1 | - | 2 | 176 | ||
EF127645.1 | - | 6 | 600 |