Gene Transcripts

Gene Symbol
Otop3
Entrez Gene ID
69602
Gene Name
otopetrin 3
Gene Aliases
2310011E08Rik
Chromosome Location
Chr. 11: 115334731 - 115346927 on Build GRCm38
UniGene ID
Mm.101844
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027132.2 NP_081408.2 6 963
GenBank mRNA AK043266.1 - 5, 6 861
BC150803.1 - 5, 6 823 9087714

View Full Product Details