Gene Transcripts

Gene Symbol
Vom1r43
Entrez Gene ID
494280
Gene Name
vomeronasal 1 receptor 43
Gene Aliases
V1rg17
Chromosome Location
Chr. 1: 67205796 - 67206713 on Build Rnor_6.0
UniGene ID
Rn.129424
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008932.1 NP_001008932.1 1 766
GenBank mRNA AY510319.1 - 1 766

View Full Product Details