Gene Transcripts

Gene Symbol
RMND1
Entrez Gene ID
55005
Gene Name
required for meiotic nuclear division 1 homolog
Gene Aliases
C6orf96, COXPD11, RMD1, bA351K16, bA351K16.3
Chromosome Location
Chr. 6: 151404548 - 151452181 on Build GRCh38
UniGene ID
Hs.486835
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271937.1 NP_001258866.1 9 820
NM_017909.3 NP_060379.2 10 1338
XM_005267040.3 XP_005267097.1 9 819
XM_017010988.1 XP_016866477.1 10 1337
GenBank mRNA AK000634.1 - 10 1281
AK026480.1 - 9 839
AK292339.1 - 10 1313
BC106065.1 - 10 1300 6165385
BC119683.1 - 10 1203 40116343

View Full Product Details