Gene Transcripts
- Gene Symbol
- GADD45G
- Entrez Gene ID
- 10912
- Gene Name
- growth arrest and DNA damage inducible gamma
- Gene Aliases
- CR6, DDIT2, GADD45gamma, GRP17
- Chromosome Location
- Chr. 9: 89605009 - 89606555 on Build GRCh38
- UniGene ID
- Hs.9701
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF079806.1 |
- |
4 |
942 |
|
View Full Product Details