Gene Transcripts

Gene Symbol
Pik3ip1
Entrez Gene ID
305472
Gene Name
phosphoinositide-3-kinase interacting protein 1
Gene Aliases
RGD1311203, zgc:66482
Chromosome Location
Chr. 14: 83559395 - 83572481 on Build Rnor_6.0
UniGene ID
Rn.138969
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017453.1 NP_001017453.1 6 1242
XM_006251293.2 XP_006251355.1 6 2451
GenBank mRNA BC092208.1 - 6 1242

View Full Product Details