Gene Transcripts

Gene Symbol
Hoxb4
Entrez Gene ID
15412
Gene Name
homeobox B4
Gene Aliases
Hox-2.6
Chromosome Location
Chr. 11: 96318267 - 96321638 on Build GRCm38
UniGene ID
Mm.3546
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010459.7 NP_034589.3 2 1274
GenBank mRNA AK133945.1 - 2 803
BC095947.1 - 2 1274 4947607
M36654.1 - 2 846

View Full Product Details