Gene Transcripts

Gene Symbol
Dock11
Entrez Gene ID
75974
Gene Name
dedicator of cytokinesis 11
Gene Aliases
5033414A21Rik, 8030476J24, Zizimin2, sph238
Chromosome Location
Chr. X: 35888627 - 36076570 on Build GRCm38
UniGene ID
Mm.32873
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009947.3 NP_001009947.2 33, 34 3771
GenBank mRNA AB116935.2 - 33, 34 3771

View Full Product Details