Gene Transcripts

Gene Symbol
Prl7b1
Entrez Gene ID
75596
Gene Name
prolactin family 7, subfamily b, member 1
Gene Aliases
1600014J19Rik, PLP-N, Prlpn
Chromosome Location
Chr. 13: 27601812 - 27610582 on Build GRCm38
UniGene ID
Mm.46087
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029355.2 NP_083631.1 6 866
GenBank mRNA AK005458.1 - 6 867

View Full Product Details