Gene Transcripts

Gene Symbol
Gstt1
Entrez Gene ID
14871
Gene Name
glutathione S-transferase, theta 1
Gene Aliases
AI255817, Gstt1-1
Chromosome Location
Chr. 10: 75783813 - 75798584 on Build GRCm38
UniGene ID
Mm.2746
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008185.3 NP_032211.3 4 555
GenBank mRNA AK002338.1 - 4 555
BC012254.1 - 4 470 4193816
BC055020.1 - 4 491 4161144
W54467.1 - 4 506
X98055.1 - 4 467

View Full Product Details